siRNA Details
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012569.2 | NP_036701.2 | 14, 15 | 1723 | ||
GenBank mRNA | M22586.1 | - | 7, 8 | 628 | ||
M65150.1 | - | 14, 15 | 1730 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012569.2 | NP_036701.2 | 14, 15 | 1723 | ||
GenBank mRNA | M22586.1 | - | 7, 8 | 628 | ||
M65150.1 | - | 14, 15 | 1730 |