siRNA Details
| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012570.2 | NP_036702.1 | 11, 12 | 1665 | ||
| GenBank mRNA | BC081841.1 | - | 11, 12 | 1549 | 7107138 | |
| FQ210745.1 | - | 1, 2 | 60 | |||
| FQ214067.1 | - | 3, 4 | 245 | |||
| U95148.1 | - | 2, 3 | 206 | |||
| X14044.1 | - | 11, 12 | 1555 | |||
| X14223.1 | - | 11, 12 | 1528 | |||

Glud1
Genomic Map