siRNA Details
| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012570.2 | NP_036702.1 | 10 | 1476 | ||
| GenBank mRNA | BC081841.1 | - | 10 | 1360 | 7107138 | |
| FQ214067.1 | - | 2 | 56 | |||
| U95148.1 | - | 1 | 17 | |||
| X14044.1 | - | 10 | 1366 | |||
| X14223.1 | - | 10 | 1339 | |||

Glud1
Genomic Map