siRNA Details
| Gene Symbol: Got1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012571.2 | NP_036703.2 | 1 | 139 | ||
| GenBank mRNA | BC061877.1 | - | 1 | 112 | 5599273 | |
| CK600227.1 | - | 1 | 158 | |||
| D00252.1 | - | 1 | 96 | |||
| FM055803.1 | - | 1 | 61 | |||
| J04171.1 | - | 1 | 80 | |||

Got1
Genomic Map