siRNA Details
| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012572.1 | NP_036704.1 | 11, 12 | 1354 | ||
| GenBank mRNA | U08257.1 | - | 11, 12 | 1354 | ||
| X59996.1 | - | 11, 12 | 1355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grik4| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012572.1 | NP_036704.1 | 11, 12 | 1354 | ||
| GenBank mRNA | U08257.1 | - | 11, 12 | 1354 | ||
| X59996.1 | - | 11, 12 | 1355 | |||