siRNA Details
| Gene Symbol: Grik5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031508.2 | NP_113696.1 | 8 | 1108 | ||
| XM_006228382.2 | XP_006228444.1 | 8 | 1106 | |||
| XM_008758889.1 | XP_008757111.1 | 7 | 1052 | |||
| GenBank mRNA | U08258.1 | - | 7 | 817 | ||
| Z11581.1 | - | 8 | 1006 | |||

Grik5
Genomic Map