siRNA Details
| Gene Symbol: Grin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270602.1 | NP_001257531.1 | 13 | 2038 | ||
| NM_001270603.1 | NP_001257532.1 | 13 | 2038 | |||
| NM_001270605.1 | NP_001257534.1 | 12 | 1975 | |||
| NM_001270606.1 | NP_001257535.1 | 13 | 2038 | |||
| NM_001270608.1 | NP_001257537.1 | 13 | 2038 | |||
| NM_001270610.1 | NP_001257539.1 | 12 | 1975 | |||
| NM_001287423.1 | NP_001274352.1 | 12 | 1975 | |||
| NM_017010.2 | NP_058706.1 | 12 | 1975 | |||
| XM_006233555.2 | XP_006233617.1 | 13 | 2044 | |||
| XM_006233556.2 | XP_006233618.1 | 12 | 1981 | |||
| GenBank mRNA | U08261.1 | - | 12 | 1886 | ||
| U08262.1 | - | 12 | 1886 | |||
| U08263.1 | - | 13 | 1925 | |||
| U08264.1 | - | 13 | 1925 | |||
| U08265.1 | - | 12 | 1886 | |||
| U08266.1 | - | 13 | 1925 | |||
| U08267.1 | - | 12 | 1886 | |||
| U08268.1 | - | 13 | 1925 | |||
| U11418.1 | - | 12 | 1791 | |||
| X63255.1 | - | 12 | 1916 | |||
| X65227.1 | - | 13 | 1714 | |||

Grin1
Genomic Map