siRNA Details
| Gene Symbol: Grin2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012575.3 | NP_036707.3 | 9 | 2118 | ||
| XM_006247706.2 | XP_006247768.1 | 7 | 1837 | |||
| XM_006247707.1 | XP_006247769.1 | 10 | 2279 | |||
| XM_006247708.2 | XP_006247770.1 | 7 | 1734 | |||
| XM_006247709.2 | XP_006247771.1 | 7 | 1734 | |||
| GenBank mRNA | D13212.1 | - | 9 | 2118 | ||
| M91563.1 | - | 7 | 1737 | |||
| U08259.1 | - | 10 | 2278 | |||

Grin2c
Genomic Map