siRNA Details
| Gene Symbol: Grin2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022797.1 | NP_073634.1 | 12 | 3378 | ||
| XM_008759316.1 | XP_008757538.1 | 11 | 3083 | |||
| XM_008759317.1 | XP_008757539.1 | 11 | 3083 | |||
| GenBank mRNA | D13213.1 | - | 11 | 2672 | ||
| L31611.1 | - | 11 | 2653 | |||
| L31612.1 | - | 12 | 3378 | |||
| U08260.1 | - | 12 | 2724 | |||

Grin2d
Genomic Map