siRNA Details
| Gene Symbol: Nr3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012576.2 | NP_036708.2 | 6 | 1985 | ||
| XM_008772065.1 | XP_008770287.1 | 6 | 1986 | |||
| XM_008772066.1 | XP_008770288.1 | 7 | 2036 | |||
| GenBank mRNA | AB115420.1 | - | 6 | 1972 | ||
| AF455050.1 | - | 6 | 1976 | |||
| AY066016.2 | - | 6 | 1980 | |||
| CB699574.1 | - | 2 | 187 | |||
| M14053.1 | - | 6 | 2043 | |||
| Y12264.1 | - | 6 | 1997 | |||

Nr3c1
Genomic Map