siRNA Details
| Gene Symbol: Nr3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012576.2 | NP_036708.2 | 4 | 1588 | ||
| XM_008772065.1 | XP_008770287.1 | 4 | 1589 | |||
| XM_008772066.1 | XP_008770288.1 | 5 | 1639 | |||
| GenBank mRNA | AB115420.1 | - | 4 | 1575 | ||
| AF455050.1 | - | 4 | 1579 | |||
| AY066016.2 | - | 4 | 1583 | |||
| M14053.1 | - | 4 | 1646 | |||
| Y12264.1 | - | 4 | 1600 | |||

Nr3c1
Genomic Map