siRNA Details
| Gene Symbol: Grm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105711.1 | NP_001099181.1 | 3 | 731 | ||
| GenBank mRNA | M92075.1 | - | 4 | 724 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grm2| Gene Symbol: Grm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105711.1 | NP_001099181.1 | 3 | 731 | ||
| GenBank mRNA | M92075.1 | - | 4 | 724 | ||
Set Membership: |
|