siRNA Details
| Gene Symbol: Gstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017014.1 | NP_058710.1 | 8 | 648 | ||
| GenBank mRNA | BC063172.1 | - | 8 | 688 | 6920688 | |
| FJ179401.1 | - | 8 | 612 | |||
| J02810.1 | - | 8 | 648 | |||
| M11719.1 | - | 8 | 649 | |||
| M28241.1 | - | 8 | 708 | |||
| X04229.1 | - | 8 | 648 | |||

Gstm1
Genomic Map