siRNA Details
| Gene Symbol: Hdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017016.2 | NP_058712.2 | 8 | 898 | ||
| GenBank mRNA | BC128724.1 | - | 8 | 898 | 8375507 | |
| M29591.1 | - | 8 | 905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdc| Gene Symbol: Hdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017016.2 | NP_058712.2 | 8 | 898 | ||
| GenBank mRNA | BC128724.1 | - | 8 | 898 | 8375507 | |
| M29591.1 | - | 8 | 905 | |||