siRNA Details
| Gene Symbol: Hmox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012580.2 | NP_036712.1 | 5 | 887 | ||
| GenBank mRNA | BC091164.1 | - | 5 | 887 | 7384448 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmox1| Gene Symbol: Hmox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012580.2 | NP_036712.1 | 5 | 887 | ||
| GenBank mRNA | BC091164.1 | - | 5 | 887 | 7384448 | |