siRNA Details
| Gene Symbol: Hp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012582.2 | NP_036714.2 | 5 | 975 | ||
| GenBank mRNA | AY325231.1 | - | 6 | 1103 | ||
| BC089816.1 | - | 5 | 975 | 7321960 | ||
| FQ209575.1 | - | 3 | 880 | |||
| FQ209592.1 | - | 5 | 1027 | |||
| FQ209684.1 | - | 5 | 1027 | |||
| FQ211144.1 | - | 5 | 1029 | |||
| FQ219122.1 | - | 5 | 1027 | |||
| FQ219413.1 | - | 2 | 785 | |||
| FQ219443.1 | - | 5 | 1028 | |||
| FQ219547.1 | - | 5 | 998 | |||
| FQ219598.1 | - | 5 | 997 | |||
| K01933.1 | - | 2 | 739 | |||

Hp
Genomic Map