siRNA Details
| Gene Symbol: Iapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012586.2 | NP_036718.1 | 2 | 178 | ||
| XM_006237614.2 | XP_006237676.1 | 3 | 707 | |||
| GenBank mRNA | J04544.1 | - | 3 | 265 | ||
| M25390.1 | - | 3 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iapp| Gene Symbol: Iapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012586.2 | NP_036718.1 | 2 | 178 | ||
| XM_006237614.2 | XP_006237676.1 | 3 | 707 | |||
| GenBank mRNA | J04544.1 | - | 3 | 265 | ||
| M25390.1 | - | 3 | 214 | |||