siRNA Details
| Gene Symbol: Igf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190162.1 | NP_001177091.1 | 4 | 1348 | ||
| NM_001190163.1 | NP_001177092.1 | 4 | 1336 | |||
| NM_031511.2 | NP_113699.2 | 4 | 2323 | |||
| XM_006230664.2 | XP_006230726.1 | 4 | 2125 | |||
| XM_006230665.2 | XP_006230727.1 | 4 | 1328 | |||
| XM_008760074.1 | XP_008758296.1 | 4 | 2146 | |||
| XM_008760075.1 | XP_008758297.1 | 5 | 1661 | |||
| GenBank mRNA | X13101.1 | - | 4 | 1361 | ||
| X14833.1 | - | 4 | 1637 | |||
| X14834.1 | - | 4 | 2392 | |||
| X16703.1 | - | 1 | 984 | |||

Igf2
Genomic Map