siRNA Details
| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012588.2 | NP_036720.2 | 3, 4 | 861 | ||
| GenBank mRNA | BF281457.1 | - | 2, 3 | 324 | ||
| M31837.1 | - | 3, 4 | 814 | |||
| M33300.1 | - | 3, 4 | 791 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp3| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012588.2 | NP_036720.2 | 3, 4 | 861 | ||
| GenBank mRNA | BF281457.1 | - | 2, 3 | 324 | ||
| M31837.1 | - | 3, 4 | 814 | |||
| M33300.1 | - | 3, 4 | 791 | |||