siRNA Details
| Gene Symbol: Il1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031512.2 | NP_113700.2 | 5 | 439 | ||
| GenBank mRNA | BC091141.1 | - | 5 | 439 | 7365029 | |
| M98820.1 | - | 5 | 459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1b| Gene Symbol: Il1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031512.2 | NP_113700.2 | 5 | 439 | ||
| GenBank mRNA | BC091141.1 | - | 5 | 439 | 7365029 | |
| M98820.1 | - | 5 | 459 | |||