siRNA Details
| Gene Symbol: Il6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012589.2 | NP_036721.1 | 4 | 501 | ||
| GenBank mRNA | M26744.1 | - | 4 | 501 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il6| Gene Symbol: Il6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012589.2 | NP_036721.1 | 4 | 501 | ||
| GenBank mRNA | M26744.1 | - | 4 | 501 | ||