siRNA Details
Gene Symbol: Ina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019128.4 | NP_062001.1 | 3 | 2767 | ||
GenBank mRNA | X52017.1 | - | 3 | 1491 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019128.4 | NP_062001.1 | 3 | 2767 | ||
GenBank mRNA | X52017.1 | - | 3 | 1491 |