siRNA Details
| Gene Symbol: Inha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012590.2 | NP_036722.1 | 2 | 1136 | ||
| GenBank mRNA | BC083564.1 | - | 2 | 1136 | 7112717 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inha| Gene Symbol: Inha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012590.2 | NP_036722.1 | 2 | 1136 | ||
| GenBank mRNA | BC083564.1 | - | 2 | 1136 | 7112717 | |