siRNA Details
| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017022.2 | NP_058718.2 | 7 | 978 | ||
| XM_006255824.1 | XP_006255886.1 | 7 | 1008 | |||
| GenBank mRNA | BC131845.1 | - | 7 | 978 | 8366766 | |
| U12309.1 | - | 7 | 852 | |||

Itgb1
Genomic Map