siRNA Details
| Gene Symbol: Kcna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173095.3 | NP_775118.1 | 1 | 644 | ||
| GenBank mRNA | M26161.1 | - | 1 | 644 | ||
| X12589.1 | - | 857 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcna1| Gene Symbol: Kcna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173095.3 | NP_775118.1 | 1 | 644 | ||
| GenBank mRNA | M26161.1 | - | 1 | 644 | ||
| X12589.1 | - | 857 | ||||