siRNA Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M64862.1 | - | 8 | 1129 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco2a1| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M64862.1 | - | 8 | 1129 | ||