siRNA Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033235.1 | NP_150238.1 | 9 | 974 | ||
| GenBank mRNA | AF093773.1 | - | 9 | 974 | ||
| BC059124.1 | - | 9 | 1015 | 6921518 | ||
| FQ212702.1 | - | 9 | 1018 | |||
| FQ214669.1 | - | 4 | 313 | |||
| FQ215129.1 | - | 9 | 1029 | |||
| FQ215603.1 | - | 9 | 1013 | |||
| FQ217819.1 | - | 1018 | ||||
| FQ224891.1 | - | 9 | 1016 | |||
| FQ233582.1 | - | 9 | 1018 | |||

Mdh1
Genomic Map