siRNA Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033235.1 | NP_150238.1 | 6 | 641 | ||
| GenBank mRNA | AF093773.1 | - | 6 | 641 | ||
| BC059124.1 | - | 6 | 682 | 6921518 | ||
| FQ212702.1 | - | 6 | 685 | |||
| FQ215129.1 | - | 6 | 696 | |||
| FQ215603.1 | - | 6 | 680 | |||
| FQ217819.1 | - | 685 | ||||
| FQ224891.1 | - | 6 | 683 | |||
| FQ233582.1 | - | 6 | 685 | |||

Mdh1
Genomic Map