siRNA Details
| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012600.2 | NP_036732.2 | 7 | 802 | ||
| XM_008766439.1 | XP_008764661.1 | 7 | 1099 | |||
| GenBank mRNA | CO560778.1 | - | 3 | 299 | ||
| FQ228526.1 | - | 7 | 833 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Me1| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012600.2 | NP_036732.2 | 7 | 802 | ||
| XM_008766439.1 | XP_008764661.1 | 7 | 1099 | |||
| GenBank mRNA | CO560778.1 | - | 3 | 299 | ||
| FQ228526.1 | - | 7 | 833 | |||