siRNA Details
| Gene Symbol: Mog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022668.2 | NP_073159.2 | 2 | 338 | ||
| XM_006255867.2 | XP_006255929.1 | 2 | 811 | |||
| XM_008772687.1 | XP_008770909.1 | 2 | 815 | |||
| GenBank mRNA | BC087717.1 | - | 2 | 338 | 7317044 | |
| FQ214199.1 | - | 2 | 318 | |||
| L21995.1 | - | 1 | 169 | |||
| M99485.1 | - | 2 | 338 | |||

Mog
Genomic Map