siRNA Details
| Gene Symbol: Mpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012601.2 | NP_036733.2 | 2 | 279 | ||
| GenBank mRNA | AW435341.1 | - | 2 | 274 | ||
| CB765983.1 | - | 1 | 26 | |||
| X56420.1 | - | 2 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpg| Gene Symbol: Mpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012601.2 | NP_036733.2 | 2 | 279 | ||
| GenBank mRNA | AW435341.1 | - | 2 | 274 | ||
| CB765983.1 | - | 1 | 26 | |||
| X56420.1 | - | 2 | 241 | |||