siRNA Details
| Gene Symbol: Muc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012602.1 | NP_036734.1 | 4 | 1648 | ||
| XM_006232590.2 | XP_006232652.1 | 5 | 1913 | |||
| XM_008761128.1 | XP_008759350.1 | 5 | 1914 | |||
| GenBank mRNA | BC166505.1 | - | 4 | 1648 | 7122817 | |

Muc1
Genomic Map