siRNA Details
| Gene Symbol: Myh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170600.1 | NP_001164071.1 | 3, 4 | 647 | ||
| XM_006245842.2 | XP_006245904.1 | 4, 5 | 754 | |||
| XM_006245844.2 | XP_006245906.1 | 4, 5 | 749 | |||
| GenBank mRNA | AY953023.1 | - | 1, 2 | 67 | ||
| BC166736.1 | - | 3, 4 | 589 | |||

Myh11
Genomic Map