siRNA Details
| Gene Symbol: Myh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012604.1 | NP_036736.1 | 28 | 3847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myh3| Gene Symbol: Myh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012604.1 | NP_036736.1 | 28 | 3847 | ||