siRNA Details
| Gene Symbol: SHBG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040.4 | NP_001031.2 | 2 | 272 | ||
| NM_001146279.2 | NP_001139751.1 | 2 | 272 | |||
| NM_001146280.2 | NP_001139752.1 | 2 | 272 | |||
| NM_001146281.2 | NP_001139753.1 | 2 | 272 | |||
| NM_001289113.1 | NP_001276042.1 | 2 | 265 | |||
| NM_001289114.1 | NP_001276043.1 | 2 | 128 | |||
| NM_001289115.1 | NP_001276044.1 | 2 | 265 | |||
| NM_001289116.1 | NP_001276045.1 | 2 | 272 | |||
| GenBank mRNA | AK302603.1 | - | 2 | 265 | ||
| BC069597.1 | - | 2 | 190 | |||
| BC101785.1 | - | 2 | 224 | 8069291 | ||
| BC112186.1 | - | 2 | 224 | 8327654 | ||
| CD013955.1 | - | 2 | 214 | |||
| CD013957.1 | - | 2 | 214 | |||
| CD013958.1 | - | 2 | 214 | |||
| EU352661.1 | - | 2 | 112 | |||
| EU352662.1 | - | 2 | 112 | |||
| EU352663.1 | - | 2 | 112 | |||
| EU352664.1 | - | 2 | 112 | |||
| EU352665.1 | - | 2 | 133 | |||
| EU352666.1 | - | 2 | 133 | |||
| EU352667.1 | - | 2 | 133 | |||
| EU352668.1 | - | 2 | 133 | |||
| EU352669.1 | - | 2 | 133 | |||
| EU352670.1 | - | 2 | 128 | |||
| EU352671.1 | - | 2 | 128 | |||
| EU352672.1 | - | 2 | 128 | |||
| EU352673.1 | - | 2 | 128 | |||
| EU352674.1 | - | 2 | 128 | |||
| EU352675.1 | - | 2 | 128 | |||
| EU352676.1 | - | 2 | 128 | |||
| X05403.1 | - | 2 | 130 | |||
| X05885.1 | - | 1 | 57 | |||
| X16351.1 | - | 2 | 178 | |||

SHBG
Genomic Map