siRNA Details
Gene Symbol: Neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031522.3 | NP_113710.2 | 3 | 679 | ||
GenBank mRNA | AB035722.1 | - | 3 | 537 | ||
BC100639.1 | - | 3 | 554 | 7929913 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031522.3 | NP_113710.2 | 3 | 679 | ||
GenBank mRNA | AB035722.1 | - | 3 | 537 | ||
BC100639.1 | - | 3 | 554 | 7929913 |