siRNA Details
| Gene Symbol: Klk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031523.1 | NP_113711.1 | 2 | 179 | ||
| GenBank mRNA | BC078784.1 | - | 2 | 161 | 7128287 | |
| J00758.1 | - | 2 | 173 | |||
| M11563.1 | - | 2 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1| Gene Symbol: Klk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031523.1 | NP_113711.1 | 2 | 179 | ||
| GenBank mRNA | BC078784.1 | - | 2 | 161 | 7128287 | |
| J00758.1 | - | 2 | 173 | |||
| M11563.1 | - | 2 | 179 | |||