siRNA Details
Gene Symbol: Nos3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021838.2 | NP_068610.1 | 8 | 1118 | ||
XM_006235872.1 | XP_006235934.1 | 8 | 1147 | |||
GenBank mRNA | AB176831.1 | - | 8 | 1118 | ||
AF110508.1 | - | 1 | 112 | |||
AY695391.1 | - | 9 | 1052 |
