siRNA Details
| Gene Symbol: Nras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080766.2 | NP_542944.1 | 5 | 573 | ||
| XM_006233059.2 | XP_006233121.1 | 5 | 617 | |||
| GenBank mRNA | BC098659.1 | - | 5 | 573 | 7368145 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nras| Gene Symbol: Nras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080766.2 | NP_542944.1 | 5 | 573 | ||
| XM_006233059.2 | XP_006233121.1 | 5 | 617 | |||
| GenBank mRNA | BC098659.1 | - | 5 | 573 | 7368145 | |