siRNA Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012619.2 | NP_036751.2 | 6 | 742 | ||
| XM_008765215.1 | XP_008763437.1 | 4 | 682 | |||
| GenBank mRNA | BC078881.1 | - | 6 | 742 | 7131414 | |
| FQ219397.1 | - | 2 | 178 | |||
| K02599.1 | - | 1 | 32 | |||
| M12337.1 | - | 6 | 727 | |||

Pah
Genomic Map