siRNA Details
| Gene Symbol: Reg3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173097.1 | NP_775120.1 | 4 | 295 | ||
| XM_008763019.1 | XP_008761241.1 | 4 | 433 | |||
| GenBank mRNA | L20869.1 | - | 4 | 295 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reg3g| Gene Symbol: Reg3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173097.1 | NP_775120.1 | 4 | 295 | ||
| XM_008763019.1 | XP_008761241.1 | 4 | 433 | |||
| GenBank mRNA | L20869.1 | - | 4 | 295 | ||