siRNA Details
| Gene Symbol: Pgam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053290.1 | NP_445742.1 | 3 | 521 | ||
| GenBank mRNA | BC065582.1 | - | 3 | 576 | 5623677 | |
| M76591.1 | - | 3 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgam1| Gene Symbol: Pgam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053290.1 | NP_445742.1 | 3 | 521 | ||
| GenBank mRNA | BC065582.1 | - | 3 | 576 | 5623677 | |
| M76591.1 | - | 3 | 521 | |||