siRNA Details
| Gene Symbol: Pgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053291.3 | NP_445743.2 | 4, 5 | 422 | ||
| GenBank mRNA | BC063161.1 | - | 4, 5 | 503 | 6921844 | |
| BC087651.1 | - | 4, 5 | 422 | 7310274 | ||
| CF976116.1 | - | 4, 5 | 521 | |||
| M31788.1 | - | 4, 5 | 458 | |||

Pgk1
Genomic Map