siRNA Details
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017033.2 | NP_058729.2 | 10 | 1612 | ||
XM_006238442.2 | XP_006238504.1 | 11 | 1803 | |||
XM_008763853.1 | XP_008762075.1 | 10 | 2155 | |||
GenBank mRNA | BC099807.1 | - | 10 | 1612 | 7937943 | |
BC128703.1 | - | 10 | 1617 | |||
FQ220427.1 | - | 3 | 402 | |||
L11694.1 | - | 10 | 1595 |
