siRNA Details
| Gene Symbol: Pla2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133551.2 | NP_598235.2 | 14, 15 | 1761 | ||
| GenBank mRNA | BC070940.1 | - | 14, 15 | 1761 | 7099813 | |
| S77829.1 | - | 14, 15 | 1747 | |||
| U38376.1 | - | 14, 15 | 1762 | |||

Pla2g4a
Genomic Map