siRNA Details
| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017035.1 | NP_058731.1 | 14 | 2332 | ||
| XM_006244063.2 | XP_006244125.1 | 13 | 2312 | |||
| GenBank mRNA | EF089258.1 | - | 13 | 2191 | ||
| M20637.1 | - | 14 | 2332 | |||

Plcd1
Genomic Map