siRNA Details
| Gene Symbol: Ppm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017038.1 | NP_058734.1 | 2 | 253 | ||
| XM_006240193.2 | XP_006240255.1 | 2 | 3556 | |||
| XM_006240194.2 | XP_006240256.1 | 2 | 510 | |||
| XM_006240195.2 | XP_006240257.1 | 2 | 274 | |||
| GenBank mRNA | J04503.1 | - | 2 | 253 | ||

Ppm1a
Genomic Map