siRNA Details
| Gene Symbol: Ppm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270619.1 | NP_001257548.1 | 4, 5 | 1486 | ||
| NM_001270620.1 | NP_001257549.1 | 5, 6 | 1960 | |||
| NM_033096.2 | NP_149087.1 | 4, 5 | 1486 | |||
| XM_008764454.1 | XP_008762676.1 | 3, 4 | 1097 | |||
| GenBank mRNA | AJ271834.1 | - | 4, 5 | 1479 | ||
| AJ271837.1 | - | 4, 5 | 1479 | |||
| BC061986.1 | - | 4, 5 | 1441 | 5599216 | ||
| BC081762.1 | - | 5, 6 | 1960 | 7113731 | ||
| CV110469.1 | - | 2, 3 | 149 | |||
| FM075941.1 | - | 2, 3 | 226 | |||
| S90449.1 | - | 4, 5 | 1772 | |||

Ppm1b
Genomic Map