siRNA Details
| Gene Symbol: Ppp1cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270983.1 | NP_001257912.1 | 2 | 408 | ||
| NM_022498.2 | NP_071943.1 | 2 | 408 | |||
| XM_008769212.1 | XP_008767434.1 | 3 | 295 | |||
| GenBank mRNA | BC078825.1 | - | 2 | 335 | 7112668 | |
| D90165.1 | - | 2 | 237 | |||
| D90166.1 | - | 2 | 351 | |||
| FN803249.1 | - | 2 | 295 | |||

Ppp1cc
Genomic Map