siRNA Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105713.1 | NP_001099183.1 | 4, 5 | 677 | ||
| XM_008768310.1 | XP_008766532.1 | 4, 5 | 914 | |||
| GenBank mRNA | BC169007.1 | - | 4, 5 | 626 | 7387040 | |
| X07286.1 | - | 4, 5 | 628 | |||

Prkca
Genomic Map